Assay Details
| Gene Symbol: LOC4346747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795965.2 | XP_015651451.1 | 3 - 4 | 388 | 82 | |
| GenBank mRNA | AK068681.1 | - | 3 - 4 | 343 | 82 | |
More Information
Set Membership: |
|

LOC4346747
Genomic Map