Assay Details
Gene Symbol: LOC4347580 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795912.2 | XP_015651398.1 | 2 - 3 | 700 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
