Assay Details
Gene Symbol: LOC4349162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015758936.2 | XP_015614422.1 | 1 - 2 | 419 | 141 | |
GenBank mRNA | AK073777.1 | - | 1 - 2 | 134 | 141 | |
AK101367.1 | - | 1 - 2 | 123 | 141 | ||
AK119563.1 | - | 1 - 2 | 134 | 141 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015758936.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
