Assay Details
Gene Symbol: LOC4349162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015758936.2 | XP_015614422.1 | 2 - 3 | 1058 | 79 | |
GenBank mRNA | AK073777.1 | - | 2 - 3 | 773 | 79 | |
AK101367.1 | - | 2 - 3 | 762 | 79 | ||
AK119563.1 | - | 2 - 3 | 773 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
