Assay Details
Gene Symbol: LOC4349162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015758936.2 | XP_015614422.1 | 3 - 4 | 2034 | 62 | |
GenBank mRNA | AK073777.1 | - | 3 - 4 | 1749 | 62 | |
AK101367.1 | - | 3 - 4 | 1738 | 62 | ||
AK119563.1 | - | 3 - 4 | 1749 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
