Assay Details
Gene Symbol: LOC4349225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015759525.2 | XP_015615011.1 | 3 - 4 | 702 | 74 | |
GenBank mRNA | AK111579.1 | - | 3 - 4 | 608 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
