Assay Details
Gene Symbol: LOC4349236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015759073.2 | XP_015614559.1 | 3 - 4 | 413 | 77 | |
GenBank mRNA | AK105227.1 | - | 3 - 4 | 375 | 77 | |
AK121221.1 | - | 3 - 4 | 421 | 77 | ||
KX434733.1 | - | 3 - 4 | 308 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
