Assay Details
Gene Symbol: LOC4349236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015759073.2 | XP_015614559.1 | 4 - 5 | 524 | 68 | |
GenBank mRNA | AK105227.1 | - | 4 - 5 | 486 | 68 | |
AK121221.1 | - | 4 - 5 | 532 | 68 | ||
KX434733.1 | - | 4 - 5 | 419 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
