Assay Details
| Gene Symbol: LOC4350215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762537.2 | XP_015618023.1 | 2 - 3 | 535 | 87 | |
| GenBank mRNA | AK065745.1 | - | 2 - 3 | 538 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015762537.2 transcripts.
Set Membership: |
|

LOC4350215
Genomic Map