Assay Details
Gene Symbol: LOC4327708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765763.1 | XP_015621249.1 | 2 - 3 | 472 | 151 | |
GenBank mRNA | AK111904.1 | - | 2 - 3 | 377 | 151 | |
AK111979.1 | - | 2 - 3 | 373 | 151 | ||
AK119548.1 | - | 2 - 3 | 352 | 151 | ||
AY620417.1 | - | 2 - 3 | 319 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
