Assay Details
Gene Symbol: LOC9266070 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774006.2 | 1 | 264 | 154 | ||
GenBank mRNA | AK108043.1 | - | 1 | 251 | 154 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015774006.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
