Assay Details
| Gene Symbol: LOC4346135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793951.2 | XP_015649437.1 | 1 | 420 | 139 | |
| XM_015793952.2 | XP_015649438.1 | 1 | 428 | 139 | ||
| GenBank mRNA | AK242980.1 | - | 1 | 429 | 139 | |
| AY177701.1 | - | 1 | 146 | 139 | ||
More Information
Set Membership: |
|

LOC4346135
Genomic Map