Assay Details
| Gene Symbol: LOC4348840 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK067249.1 | - | 3 - 4 | 1078 | 144 | |
| JF969232.1 | - | 3 - 4 | 1045 | 144 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4348840| Gene Symbol: LOC4348840 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK067249.1 | - | 3 - 4 | 1078 | 144 | |
| JF969232.1 | - | 3 - 4 | 1045 | 144 | ||
Set Membership: |
|