Assay Details
Gene Symbol: LOC4324360 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765443.1 | XP_015620929.1 | 3 | 582 | 127 | |
GenBank mRNA | AK058869.1 | - | 3 | 545 | 127 | |
AK069668.1 | - | 1 | 1872 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
