Assay Details
Gene Symbol: LOC4331194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770086.2 | XP_015625572.1 | 2 | 1250 | 78 | |
GenBank mRNA | AK102174.1 | - | 2 | 1257 | 78 | |
BT130343.1 | - | 599 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
