Assay Details
| Gene Symbol: LOC4335343 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778267.2 | XP_015633753.1 | 10 | 1355 | 131 | |
| XM_015778268.2 | XP_015633754.1 | 10 | 1055 | 131 | ||
| GenBank mRNA | AK060035.1 | - | 9 | 851 | 131 | |
More Information
Set Membership: |
|

LOC4335343
Genomic Map