Assay Details
Gene Symbol: LOC4336500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780748.2 | XP_015636234.1 | 1 | 76 | 112 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015780748.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
