Assay Details
Gene Symbol: LOC4339324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782161.2 | XP_015637647.1 | 2 | 868 | 153 | |
GenBank mRNA | AK060246.1 | - | 744 | 153 | ||
AK061969.1 | - | 2 | 753 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
