Assay Details
| Gene Symbol: GRK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141515.2 | XP_014997001.1 | 892 | 69 | ||
| XM_015141517.2 | XP_014997003.1 | 890 | 69 | |||
| XM_015141518.2 | XP_014997004.1 | 1593 | 69 | |||
| XM_028849953.1 | XP_028705786.1 | 720 | 69 | |||
| GenBank mRNA | JU327344.1 | - | 717 | 69 | ||
| JU470798.1 | - | 900 | 69 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GRK6
Genomic Map