Assay Details
| Gene Symbol: ASZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168607.1 | NP_001162078.1 | 714 | 100 | ||
| XM_015134749.2 | XP_014990235.1 | 348 | 100 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASZ1| Gene Symbol: ASZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168607.1 | NP_001162078.1 | 714 | 100 | ||
| XM_015134749.2 | XP_014990235.1 | 348 | 100 | |||
Set Membership: |
|