Assay Details
| Gene Symbol: TM4SF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194166.2 | NP_001181095.2 | 282 | 91 | ||
| XM_028843334.1 | XP_028699167.1 | 588 | 91 | |||
| XM_028843335.1 | XP_028699168.1 | 588 | 91 | |||
More Information
Set Membership: |
|

TM4SF19
Genomic Map