Assay Details
Gene Symbol: WTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260846.1 | NP_001247775.1 | 677 | 75 | ||
XM_015137269.2 | XP_014992755.2 | 687 | 75 | |||
GenBank mRNA | JU322249.1 | - | 657 | 75 | ||
JU472926.1 | - | 671 | 75 | |||
JV043734.1 | - | 677 | 75 | |||
JV638425.1 | - | 681 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
