Assay Details
Gene Symbol: PIP4K2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261154.1 | NP_001248083.1 | 448 | 85 | ||
GenBank mRNA | JU336433.1 | - | 440 | 85 | ||
JU477249.1 | - | 417 | 85 | |||
JV048280.1 | - | 448 | 85 | |||
JV602242.1 | - | 497 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
