Assay Details
Gene Symbol: HJV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015149644.2 | XP_015005130.2 | 340 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015149644.2 transcripts.
Set Membership: |
![]() ![]() |
