Assay Details
Gene Symbol: ZKSCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194124.2 | NP_001181053.2 | 814 | 82 | ||
XM_028844987.1 | XP_028700820.1 | 814 | 82 | |||
GenBank mRNA | JU336283.1 | - | 795 | 82 | ||
JU336284.1 | - | 795 | 82 | |||
JU336285.1 | - | 795 | 82 | |||
JU472805.1 | - | 790 | 82 | |||
JV048195.1 | - | 790 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
