Assay Details
Gene Symbol: ZNF184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015135754.2 | XP_014991240.2 | 613 | 74 | ||
XM_015135755.2 | XP_014991241.2 | 817 | 74 | |||
XM_015135756.2 | XP_014991242.2 | 880 | 74 | |||
XM_015135757.2 | XP_014991243.2 | 477 | 74 | |||
XM_028847766.1 | XP_028703599.1 | 598 | 74 | |||
XM_028847767.1 | XP_028703600.1 | 666 | 74 | |||
GenBank mRNA | JU336302.1 | - | 556 | 74 | ||
JU477134.1 | - | 583 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015135757.2, XM_028847766.1, XM_028847767.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
