Assay Details
Gene Symbol: ERICH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107825.4 | XP_001107825.2 | 1130 | 64 | ||
XM_015131802.2 | XP_014987288.2 | 1033 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
