Assay Details
| Gene Symbol: TMCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266303.1 | NP_001253232.1 | 2267 | 86 | ||
| XM_015118391.2 | XP_014973877.1 | 2165 | 86 | |||
| XM_015118397.2 | XP_014973883.1 | 1356 | 86 | |||
| XM_015118406.2 | XP_014973892.1 | 1266 | 86 | |||
| XM_028832082.1 | XP_028687915.1 | 1273 | 86 | |||
| GenBank mRNA | JU337383.1 | - | 2267 | 86 | ||
| JV599976.1 | - | 2314 | 86 | |||
| JV702997.1 | - | 2300 | 86 | |||
More Information
Set Membership: |
|

TMCC2
Genomic Map