Assay Details
| Gene Symbol: CDYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126680.2 | XP_014982166.1 | 955 | 60 | ||
| XM_028841318.1 | XP_028697151.1 | 1015 | 60 | |||
| GenBank mRNA | JV641991.1 | - | 876 | 60 | ||
| JV704125.1 | - | 894 | 60 | |||
More Information
Set Membership: |
|

CDYL2
Genomic Map