Assay Details
| Gene Symbol: REM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100692.4 | XP_001100692.1 | 637 | 115 | ||
| GenBank mRNA | JV708173.1 | - | 560 | 115 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

REM2| Gene Symbol: REM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100692.4 | XP_001100692.1 | 637 | 115 | ||
| GenBank mRNA | JV708173.1 | - | 560 | 115 | ||
Set Membership: |
|