Assay Details
Gene Symbol: WASHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015152660.2 | XP_015008146.1 | 2411 | 99 | ||
GenBank mRNA | JU320457.1 | - | 2388 | 99 | ||
JU471911.1 | - | 2357 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
