Assay Details
| Gene Symbol: SYNPO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139136.2 | XP_014994622.2 | 280 | 87 | ||
| XM_015139137.2 | XP_014994623.2 | 280 | 87 | |||
| XM_015139138.2 | XP_014994624.1 | 281 | 87 | |||
| GenBank mRNA | JV043350.1 | - | 255 | 87 | ||
| JV043351.1 | - | 255 | 87 | |||
| JV635461.1 | - | 256 | 87 | |||
| JV636970.1 | - | 256 | 87 | |||
More Information
Set Membership: |
|

SYNPO2
Genomic Map