Assay Details
| Gene Symbol: HACD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194139.3 | NP_001181068.1 | 656 | 119 | ||
| XM_015117612.2 | XP_014973098.1 | 1021 | 119 | |||
| XM_028834356.1 | XP_028690189.1 | 730 | 119 | |||
| XM_028834358.1 | XP_028690191.1 | 1195 | 119 | |||
| GenBank mRNA | JU475584.1 | - | 655 | 119 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HACD4
Genomic Map