Assay Details
| Gene Symbol: ZPBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119546.1 | XP_014975032.1 | 819 | 89 | ||
| XM_015119548.2 | XP_014975034.1 | 819 | 89 | |||
| XM_028836452.1 | XP_028692285.1 | 714 | 89 | |||
More Information
Set Membership: |
|

ZPBP2
Genomic Map