Assay Details
| Gene Symbol: C7H15orf61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194312.2 | NP_001181241.1 | 399 | 74 | ||
| GenBank mRNA | JV044901.1 | - | 412 | 74 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

C7H15orf61
Genomic Map