Assay Details
| Gene Symbol: WDR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116420.2 | XP_014971906.2 | 812 | 65 | ||
| XM_015116421.2 | XP_014971907.1 | 902 | 65 | |||
| XM_015116422.2 | XP_014971908.1 | 838 | 65 | |||
| XM_015116423.2 | XP_014971909.1 | 869 | 65 | |||
| XM_028834646.1 | XP_028690479.1 | 1270 | 65 | |||
| GenBank mRNA | JU320526.1 | - | 792 | 65 | ||
| JU471799.1 | - | 752 | 65 | |||
More Information
Set Membership: |
|

WDR5
Genomic Map