Assay Details
| Gene Symbol: NR2F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140413.2 | XP_014995899.1 | 2492 | 71 | ||
| GenBank mRNA | JU328392.1 | - | 345 | 71 | ||
| JV627193.1 | - | 527 | 71 | |||
| JV667463.1 | - | 559 | 71 | |||
More Information
Set Membership: |
|

NR2F1
Genomic Map