Assay Details
Gene Symbol: ARHGAP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839122.1 | XP_028694955.1 | 4348 | 91 | ||
GenBank mRNA | JU334158.1 | - | 3830 | 91 | ||
JU475285.1 | - | 3837 | 91 | |||
JV046843.1 | - | 3835 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
