Assay Details
| Gene Symbol: CAMK2N2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193366.2 | NP_001180295.2 | 167 | 90 | ||
| GenBank mRNA | JV565867.1 | - | 340 | 90 | ||
| JV601721.1 | - | 393 | 90 | |||
More Information
Set Membership: |
|

CAMK2N2
Genomic Map