Assay Details
| Gene Symbol: MTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190917.2 | NP_001177846.1 | 674 | 80 | ||
| XM_015148496.2 | XP_015003982.1 | 1100 | 80 | |||
| GenBank mRNA | JU337094.1 | - | 701 | 80 | ||
| JU477828.1 | - | 735 | 80 | |||
| JV048688.1 | - | 730 | 80 | |||
| JV487012.1 | - | 630 | 80 | |||
| JV598344.1 | - | 730 | 80 | |||
More Information
Set Membership: |
|

MTG1
Genomic Map