Assay Details
Gene Symbol: CDK5RAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261645.1 | NP_001248574.1 | 129 | 59 | ||
XM_015119373.2 | XP_014974859.1 | 217 | 59 | |||
XM_028835474.1 | XP_028691307.1 | 141 | 59 | |||
GenBank mRNA | JU322715.1 | - | 80 | 59 | ||
JU472133.1 | - | 129 | 59 | |||
JV045101.1 | - | 94 | 59 | |||
JV668832.1 | - | 128 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
