Assay Details
| Gene Symbol: IGSF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257906.1 | NP_001244835.1 | 826 | 104 | ||
| XM_028843355.1 | XP_028699188.1 | 978 | 104 | |||
| XM_028843356.1 | XP_028699189.1 | 728 | 104 | |||
| GenBank mRNA | JU322970.1 | - | 826 | 104 | ||
| JV044976.1 | - | 827 | 104 | |||
More Information
Set Membership: |
|

IGSF11
Genomic Map