Assay Details
Gene Symbol: C4H6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258161.2 | NP_001245090.1 | 746 | 93 | ||
XM_015136150.2 | XP_014991636.1 | 865 | 93 | |||
XM_028846875.1 | XP_028702708.1 | 1271 | 93 | |||
XM_028846876.1 | XP_028702709.1 | 1052 | 93 | |||
XM_028846877.1 | XP_028702710.1 | 1020 | 93 | |||
XM_028846879.1 | XP_028702712.1 | 1059 | 93 | |||
XM_028846880.1 | XP_028702713.1 | 1183 | 93 | |||
XM_028846881.1 | XP_028702714.1 | 933 | 93 | |||
XM_028846882.1 | XP_028702715.1 | 864 | 93 | |||
XM_028846883.1 | XP_028702716.1 | 953 | 93 | |||
XM_028846884.1 | XP_028702717.1 | 872 | 93 | |||
XM_028846885.1 | XP_028702718.1 | 979 | 93 | |||
XM_028846886.1 | XP_028702719.1 | 573 | 93 | |||
XM_028846887.1 | XP_028702720.1 | 852 | 93 | |||
XM_028846888.1 | XP_028702721.1 | 721 | 93 | |||
GenBank mRNA | JU474691.1 | - | 746 | 93 | ||
JV536770.1 | - | 574 | 93 | |||
JV635401.1 | - | 638 | 93 | |||
JV676097.1 | - | 711 | 93 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
