Assay Details
Gene Symbol: KCTD20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257899.1 | NP_001244828.1 | 1019 | 119 | ||
XM_015136134.2 | XP_014991620.1 | 720 | 119 | |||
XM_028846873.1 | XP_028702706.1 | 1106 | 119 | |||
XM_028846874.1 | XP_028702707.1 | 577 | 119 | |||
GenBank mRNA | JU333420.1 | - | 1019 | 119 | ||
JU474701.1 | - | 946 | 119 | |||
JV046459.1 | - | 1005 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
