Assay Details
Gene Symbol: RSPO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015135676.2 | XP_014991162.1 | 2201 | 118 | ||
XM_028835842.1 | XP_028691675.1 | 2240 | 118 | |||
XM_028835844.1 | XP_028691677.1 | 1101 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
