Assay Details
| Gene Symbol: UBR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257430.1 | NP_001244359.1 | 386 | 124 | ||
| XM_015144315.2 | XP_014999801.1 | 573 | 124 | |||
| XM_015144316.2 | XP_014999802.1 | 439 | 124 | |||
| XM_028850245.1 | XP_028706078.1 | 573 | 124 | |||
| GenBank mRNA | JU335244.1 | - | 386 | 124 | ||
| JV717960.1 | - | 279 | 124 | |||
More Information
Set Membership: |
|

UBR7
Genomic Map