Assay Details
Gene Symbol: ZNF207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244630.1 | NP_001231559.1 | 651 | 61 | ||
GenBank mRNA | CN648226.1 | - | 575 | 61 | ||
JU336311.1 | - | 616 | 61 | |||
JU336313.1 | - | 616 | 61 | |||
JU470793.1 | - | 617 | 61 | |||
JU470794.1 | - | 617 | 61 | |||
JU477140.1 | - | 617 | 61 | |||
JV043213.1 | - | 649 | 61 | |||
JV043214.1 | - | 649 | 61 | |||
JV043215.1 | - | 649 | 61 | |||
JV593099.1 | - | 617 | 61 | |||
JV660957.1 | - | 617 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
