Assay Details
Gene Symbol: ZSWIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015138173.2 | XP_014993659.2 | 2722 | 61 | ||
GenBank mRNA | JU327411.1 | - | 1908 | 61 | ||
JV719883.1 | - | 1897 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
