Assay Details
| Gene Symbol: SARNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151925.2 | XP_015007411.1 | 458 | 76 | ||
| XM_015151926.2 | XP_015007412.1 | 458 | 76 | |||
| XM_015151928.2 | XP_015007414.1 | 458 | 76 | |||
| XM_028829586.1 | XP_028685419.1 | 458 | 76 | |||
| GenBank mRNA | JU320935.1 | - | 398 | 76 | ||
| JU336712.1 | - | 389 | 76 | |||
| JV045519.1 | - | 420 | 76 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SARNP
Genomic Map