Assay Details
Gene Symbol: SPOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258173.2 | NP_001245102.1 | 712 | 71 | ||
XM_015119237.1 | XP_014974723.1 | 821 | 71 | |||
XM_015119238.2 | XP_014974724.1 | 1004 | 71 | |||
XM_015119242.2 | XP_014974728.1 | 872 | 71 | |||
XM_028835498.1 | XP_028691331.1 | 878 | 71 | |||
XM_028835499.1 | XP_028691332.1 | 562 | 71 | |||
XM_028835500.1 | XP_028691333.1 | 635 | 71 | |||
GenBank mRNA | JU320286.1 | - | 676 | 71 | ||
JU472235.1 | - | 768 | 71 | |||
JU472236.1 | - | 717 | 71 | |||
JV048411.1 | - | 688 | 71 | |||
JV636754.1 | - | 768 | 71 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
